Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pprc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081214.1 | NP_001074683.1 | 3 - 4 | 527 | 90 | |
GenBank mRNA | AK044522.1 | - | 3 - 4 | 486 | 90 | |
AK122322.1 | - | 3 - 4 | 485 | 90 | ||
AK147947.1 | - | 3 - 4 | 508 | 90 | ||
AK191373.1 | - | 2 - 3 | 330 | 90 | ||
BC066048.1 | - | 3 - 4 | 481 | 6825158 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |