Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081409.1 | NP_001074878.1 | 16 - 17 | 2406 | 81 | |
GenBank mRNA | AK040888.1 | - | 7 - 8 | 1137 | 81 | |
AK053606.1 | - | 6 - 7 | 765 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |