Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033461.3 | NP_001028633.1 | 4 - 5 | 658 | 62 | |
NM_001289821.1 | NP_001276750.1 | 4 - 5 | 658 | 62 | ||
NM_001289822.1 | NP_001276751.1 | 4 - 5 | 658 | 62 | ||
GenBank mRNA | AK133051.1 | - | 4 - 5 | 659 | 62 | |
AK167318.1 | - | 4 - 5 | 657 | 62 | ||
AK167729.1 | - | 4 - 5 | 657 | 62 | ||
BC150919.1 | - | 4 - 5 | 672 | 9087830 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |