Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145841.2 | NP_665840.2 | 3 - 4 | 1082 | 129 | |
GenBank mRNA | AK048960.1 | - | 3 - 4 | 1046 | 129 | |
AK136779.1 | - | 3 - 4 | 1083 | 129 | ||
AY095374.1 | - | 2 - 3 | 298 | 129 | ||
AY250713.1 | - | 2 - 3 | 298 | 129 | ||
BC104048.1 | - | 2 - 3 | 298 | 40040143 | 129 | |
BC104049.2 | - | 2 - 3 | 298 | 40040144 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |