Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033481.1 | NP_001028653.1 | 2 - 3 | 277 | 65 | |
GenBank mRNA | AK031485.1 | - | 2 - 3 | 280 | 65 | |
AK141695.1 | - | 2 - 3 | 278 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |