Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026748.2 | NP_081024.3 | 1 - 2 | 348 | 140 | |
GenBank mRNA | AB257854.1 | - | 1 - 2 | 308 | 140 | |
AK155838.1 | - | 1 - 2 | 344 | 140 | ||
BC063266.1 | - | 1 - 2 | 323 | 6409930 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |