Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026748.2 | NP_081024.3 | 37 - 38 | 5684 | 86 | |
GenBank mRNA | AB257854.1 | - | 37 - 38 | 5644 | 86 | |
AK148183.1 | - | 8 - 9 | 1140 | 86 | ||
BC010333.1 | - | 8 - 9 | 1265 | 86 | ||
BC063266.1 | - | 36 - 37 | 5452 | 6409930 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |