Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027902.2 | NP_082178.2 | 9 - 10 | 1324 | 85 | |
GenBank mRNA | AK004939.2 | - | 9 - 10 | 1171 | 85 | |
AY240929.1 | - | 9 - 10 | 1301 | 85 | ||
BC029645.1 | - | 9 - 10 | 1301 | 4195486 | 85 | |
BC057674.1 | - | 9 - 10 | 1266 | 4162007 | 85 | |
EU190436.1 | - | 8 - 9 | 1115 | 85 | ||
EU190437.1 | - | 8 - 9 | 1115 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |