Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008368.4 | NP_032394.1 | 7 - 8 | 852 | 89 | |
GenBank mRNA | AK017288.1 | - | 7 - 8 | 852 | 89 | |
AK035289.1 | - | 7 - 8 | 805 | 89 | ||
AK157572.1 | - | 7 - 8 | 853 | 89 | ||
AK169481.1 | - | 7 - 8 | 833 | 89 | ||
BC026869.1 | - | 7 - 8 | 825 | 4015306 | 89 | |
BC099616.1 | - | 7 - 8 | 826 | 30024496 | 89 | |
M28052.1 | - | 7 - 8 | 809 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |