Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021392.4 | NP_067367.3 | 5 - 6 | 639 | 60 | |
GenBank mRNA | AF242858.1 | - | 5 - 6 | 507 | 60 | |
AK031638.1 | - | 5 - 6 | 642 | 60 | ||
BC011174.1 | - | 5 - 6 | 524 | 4207033 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |