Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Radil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178702.4 | NP_848817.2 | 11 - 12 | 2661 | 76 | |
GenBank mRNA | AK052964.1 | - | 11 - 12 | 2572 | 76 | |
AK079539.1 | - | 11 - 12 | 2096 | 76 | ||
AK083410.1 | - | 11 - 12 | 2679 | 76 | ||
AK154169.1 | - | 11 - 12 | 2606 | 76 | ||
AK173277.1 | - | 11 - 12 | 2625 | 76 | ||
BC056483.1 | - | 11 - 12 | 2613 | 5696312 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |