Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvalb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013645.3 | NP_038673.2 | 2 - 3 | 216 | 113 | |
GenBank mRNA | AK002354.1 | - | 2 - 3 | 224 | 113 | |
AK005371.1 | - | 2 - 3 | 214 | 113 | ||
AK013561.1 | - | 2 - 3 | 208 | 113 | ||
AK131986.1 | - | 2 - 3 | 286 | 113 | ||
BC027424.1 | - | 2 - 3 | 216 | 4925213 | 113 | |
S75909.1 | - | 3 - 4 | 277 | 113 | ||
S75910.1 | - | 1153 | 113 | |||
X54613.1 | - | 2 - 3 | 194 | 113 | ||
X59382.1 | - | 3 - 4 | 250 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |