Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008677.2 | NP_032703.2 | 5 - 6 | 714 | 123 | |
GenBank mRNA | AB002665.1 | - | 5 - 6 | 691 | 123 | |
AK150510.1 | - | 5 - 6 | 696 | 123 | ||
AK170854.1 | - | 2 - 3 | 222 | 123 | ||
AK171315.1 | - | 5 - 6 | 492 | 123 | ||
BC025517.1 | - | 5 - 6 | 543 | 5321182 | 123 | |
FJ168470.1 | - | 5 - 6 | 505 | 123 | ||
U59488.1 | - | 5 - 6 | 661 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |