Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139646 |
BC145767.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139646 |
BC117809.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139646 |
BC129974.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139646 |
BC138548.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P139646 |
BC054740.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577001 |
AK038794.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577001 |
AK047979.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577001 |
AK077030.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577001 |
BC092219.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577001 |
NM_175751.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121648 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446452 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
56131 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633262 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
747394 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481750 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
85620 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240000 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703389 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13441 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
884284 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178298 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66229 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
33695 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P106026 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146130 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139646 |
BC145767.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139646 |
BC117809.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139646 |
BC129974.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139646 |
BC138548.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P139646 |
BC054740.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P243075 |
BC145767.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P243075 |
BC117809.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P243075 |
BC129974.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P243075 |
BC138548.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P243075 |
BC054740.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P17207 |
BC054740.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P17207 |
AK145339.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P17207 |
NM_001168504.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P17207 |
NM_001004762.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975309 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975301 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975297 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975298 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975311 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975313 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975300 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975318 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975304 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975302 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975307 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975323 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975320 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975315 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975305 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24975316 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P106026 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P146130 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06126464 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06126506 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P243075 |
BC145767.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243075 |
BC117809.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243075 |
BC129974.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243075 |
BC138548.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243075 |
BC054740.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P17207 |
BC054740.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P17207 |
AK145339.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P17207 |
NM_001168504.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P17207 |
NM_001004762.3 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P106026 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P146130 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06147964 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06147959 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06147962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975462 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975362 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975332 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975314 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975333 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24975468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975336 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975315 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975398 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975316 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05943533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975284 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06272708 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06603279 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05943538 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975332 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975304 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975407 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975299 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975307 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975292 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975288 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975343 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06474881 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975302 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975333 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975339 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975314 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975291 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06547475 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975286 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975294 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975405 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975309 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05943530 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975323 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975289 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06272700 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975318 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975401 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975290 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975297 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06547473 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06260739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975345 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975342 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06303842 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975320 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975311 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05934014 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24975313 |
- |
- |