Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oosp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033283.2 | NP_001028455.2 | 3 - 4 | 392 | 83 | |
GenBank mRNA | AK143410.1 | - | 3 - 4 | 393 | 83 | |
BC094934.1 | - | 3 - 4 | 354 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |