Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081319.1 | NP_001074788.1 | 7 - 8 | 1053 | 70 | |
GenBank mRNA | AK156653.1 | - | 7 - 8 | 954 | 70 | |
AK173171.1 | - | 6 - 7 | 635 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |