Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081213.1 | NP_001074682.1 | 2 - 3 | 687 | 81 | |
GenBank mRNA | AK041066.1 | - | 2 - 3 | 658 | 81 | |
AK050284.1 | - | 2 - 3 | 683 | 81 | ||
AK081674.1 | - | 1 - 2 | 204 | 81 | ||
AK129447.1 | - | 2 - 3 | 615 | 81 | ||
AK137206.1 | - | 2 - 3 | 643 | 81 | ||
BC086627.1 | - | 2 - 3 | 628 | 81 | ||
BC151181.1 | - | 2 - 3 | 646 | 9088070 | 81 | |
BC151182.1 | - | 2 - 3 | 646 | 9088071 | 81 | |
BC157914.1 | - | 2 - 3 | 646 | 9088152 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |