Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081213.1 | NP_001074682.1 | 5 - 6 | 1066 | 109 | |
GenBank mRNA | AK041066.1 | - | 5 - 6 | 1037 | 109 | |
AK050284.1 | - | 5 - 6 | 1062 | 109 | ||
AK081674.1 | - | 4 - 5 | 583 | 109 | ||
AK129447.1 | - | 5 - 6 | 994 | 109 | ||
AK137206.1 | - | 5 - 6 | 1022 | 109 | ||
BC086627.1 | - | 5 - 6 | 1007 | 109 | ||
BC151181.1 | - | 5 - 6 | 1025 | 9088070 | 109 | |
BC151182.1 | - | 5 - 6 | 1025 | 9088071 | 109 | |
BC157914.1 | - | 5 - 6 | 1025 | 9088152 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |