Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081308.2 | NP_001074777.1 | 3 - 4 | 467 | 92 | |
NM_001199685.1 | NP_001186614.1 | 4 - 5 | 575 | 92 | ||
NM_183306.2 | NP_899129.2 | 4 - 5 | 552 | 92 | ||
GenBank mRNA | AK040531.1 | - | 4 - 5 | 576 | 92 | |
AK042181.1 | - | 4 - 5 | 475 | 92 | ||
AK155208.1 | - | 3 - 4 | 462 | 92 | ||
AK171353.1 | - | 3 - 4 | 442 | 92 | ||
BC158072.1 | - | 4 - 5 | 517 | 9088167 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |