Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159650.1 | NP_001153122.1 | 3 - 4 | 256 | 72 | |
NM_021352.3 | NP_067327.1 | 3 - 4 | 326 | 72 | ||
GenBank mRNA | AJ272229.2 | - | 2 - 3 | 310 | 72 | |
AK053552.1 | - | 1 - 2 | 640 | 72 | ||
BC031442.1 | - | 2 - 3 | 317 | 4501690 | 72 | |
BC058522.1 | - | 2 - 3 | 235 | 6824229 | 72 | |
CJ046471.1 | - | 3 - 4 | 327 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |