Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ide | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031156.3 | NP_112419.3 | 14 - 15 | 1910 | 94 | |
GenBank mRNA | AJ278422.1 | - | 14 - 15 | 1930 | 94 | |
AK128986.1 | - | 14 - 15 | 1927 | 94 | ||
BC041675.1 | - | 14 - 15 | 1942 | 4951971 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |