Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079873.1 | NP_001073342.1 | 5 - 6 | 748 | 99 | |
NM_054054.2 | NP_473395.2 | 5 - 6 | 748 | 99 | ||
GenBank mRNA | AB208640.1 | - | 3 - 4 | 441 | 99 | |
AF358660.1 | - | 5 - 6 | 783 | 99 | ||
AK077137.1 | - | 5 - 6 | 768 | 99 | ||
BY726053.1 | - | 1 - 2 | 74 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |