Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285806.1 | NP_001272735.1 | 9 - 10 | 838 | 69 | |
NM_177078.4 | NP_796052.2 | 9 - 10 | 799 | 69 | ||
GenBank mRNA | AK076533.1 | - | 9 - 10 | 697 | 69 | |
AK134672.1 | - | 9 - 10 | 776 | 69 | ||
AK141186.1 | - | 9 - 10 | 817 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |