Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009745.2 | NP_033875.2 | 2 - 3 | 303 | 65 | |
GenBank mRNA | AJ011145.1 | - | 2 - 3 | 183 | 65 | |
AK131643.1 | - | 2 - 3 | 199 | 65 | ||
AK155076.1 | - | 2 - 3 | 185 | 65 | ||
BC011471.1 | - | 2 - 3 | 280 | 3660754 | 65 | |
BY087167.1 | - | 2 - 3 | 303 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |