Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009879.3 | NP_034009.2 | 5 - 6 | 628 | 91 | |
GenBank mRNA | AF354757.1 | - | 5 - 6 | 628 | 91 | |
AK013424.1 | - | 5 - 6 | 622 | 91 | ||
AK161375.1 | - | 5 - 6 | 617 | 91 | ||
BC055721.1 | - | 5 - 6 | 585 | 6810406 | 91 | |
Y10495.2 | - | 5 - 6 | 628 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |