Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016681.3 | NP_057890.1 | 14 - 15 | 132 | ||
GenBank mRNA | AF086905.1 | - | 13 - 14 | 132 | ||
AK044913.1 | - | 14 - 15 | 132 | |||
AK170235.1 | - | 13 - 14 | 1536 | 132 | ||
BC056617.1 | - | 14 - 15 | 4977960 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |