Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbrsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142642.1 | NP_001136114.1 | 2 - 3 | 824 | 92 | |
GenBank mRNA | BC059932.1 | - | 2 - 3 | 807 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |