Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039153.3 | NP_001034242.2 | 12 - 13 | 1609 | 96 | |
NR_104441.1 | 11 - 12 | 1532 | 96 | |||
GenBank mRNA | AK036568.1 | - | 12 - 13 | 1591 | 96 | |
AK138661.1 | - | 11 - 12 | 1531 | 96 | ||
AK144772.1 | - | 12 - 13 | 1585 | 96 | ||
DQ278867.1 | - | 12 - 13 | 1610 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |