Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164573.1 | NP_001158045.1 | 27 - 28 | 2778 | 114 | |
GenBank mRNA | AK014505.1 | - | 6 - 7 | 708 | 114 | |
BC144867.1 | - | 6 - 7 | 683 | 9053415 | 114 | |
BC144868.1 | - | 6 - 7 | 684 | 114 | ||
BC144869.1 | - | 6 - 7 | 611 | 114 | ||
BC144870.1 | - | 6 - 7 | 659 | 9053418 | 114 | |
BC144872.1 | - | 5 - 6 | 627 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |