Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122730.2 | NP_001116202.2 | 4 - 5 | 2306 | 65 | |
NM_178242.2 | NP_839973.1 | 4 - 5 | 2306 | 65 | ||
GenBank mRNA | AK141849.1 | - | 1 - 2 | 1398 | 65 | |
AK173280.1 | - | 1 - 2 | 61 | 65 | ||
BC049818.1 | - | 4 - 5 | 2306 | 6511384 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |