Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172276.3 | NP_758480.2 | 4 - 5 | 759 | 64 | |
GenBank mRNA | AK031872.1 | - | 4 - 5 | 750 | 64 | |
AK088182.1 | - | 4 - 5 | 703 | 64 | ||
AK140369.1 | - | 4 - 5 | 752 | 64 | ||
AK212067.1 | - | 2 - 3 | 190 | 64 | ||
BC057314.1 | - | 4 - 5 | 2689 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |