Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172276.3 | NP_758480.2 | 6 - 7 | 1087 | 51 | |
GenBank mRNA | AK031872.1 | - | 6 - 7 | 1078 | 51 | |
AK088182.1 | - | 6 - 7 | 1031 | 51 | ||
AK140369.1 | - | 6 - 7 | 1080 | 51 | ||
BC023268.1 | - | 1 - 2 | 118 | 51 | ||
BC057314.1 | - | 6 - 7 | 3017 | 51 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |