Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289507.1 | NP_001276436.1 | 11 - 12 | 1655 | 92 | |
NM_001289509.1 | NP_001276438.1 | 13 - 14 | 1674 | 92 | ||
NM_001289511.1 | NP_001276440.1 | 12 - 13 | 1532 | 92 | ||
NM_015799.4 | NP_056614.3 | 13 - 14 | 1614 | 92 | ||
GenBank mRNA | AF207741.1 | - | 11 - 12 | 1489 | 92 | |
AF222895.1 | - | 12 - 13 | 1532 | 92 | ||
AK004848.2 | - | 13 - 14 | 1590 | 92 | ||
AK004965.1 | - | 13 - 14 | 1645 | 92 | ||
AK155526.1 | - | 12 - 13 | 1752 | 92 | ||
AK165320.1 | - | 11 - 12 | 1483 | 92 | ||
BC013654.1 | - | 11 - 12 | 1655 | 4196597 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |