Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008052.3 | NP_032078.2 | 7 - 8 | 2255 | 68 | |
GenBank mRNA | AB015422.1 | - | 6 - 7 | 1858 | 68 | |
AK088630.1 | - | 7 - 8 | 2211 | 68 | ||
AK169455.1 | - | 7 - 8 | 2256 | 68 | ||
AK220514.1 | - | 6 - 7 | 1836 | 68 | ||
BC053055.1 | - | 7 - 8 | 1401 | 5685525 | 68 | |
BC065075.1 | - | 8 - 9 | 2037 | 68 | ||
U38252.1 | - | 5 - 6 | 1440 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |