Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB112924.1 | - | 12 - 13 | 1186 | 71 | |
AK139040.1 | - | 11 - 12 | 1129 | 71 | ||
BC042585.1 | - | 12 - 13 | 1353 | 5364086 | 71 | |
BC050883.1 | - | 12 - 13 | 1320 | 5698459 | 71 | |
D29965.1 | - | 12 - 13 | 1163 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |