Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB112924.1 | - | 5 - 6 | 468 | 87 | |
AK139040.1 | - | 4 - 5 | 410 | 87 | ||
BC042585.1 | - | 5 - 6 | 635 | 5364086 | 87 | |
BC050883.1 | - | 5 - 6 | 602 | 5698459 | 87 | |
D29965.1 | - | 5 - 6 | 445 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |