Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033343.1 | NP_001028515.1 | 3 - 4 | 298 | 77 | |
GenBank mRNA | BB626493.1 | - | 3 - 4 | 299 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |