Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145499.2 | NP_663474.2 | 6 - 7 | 982 | 83 | |
GenBank mRNA | AK050082.1 | - | 6 - 7 | 983 | 83 | |
AY227736.1 | - | 6 - 7 | 966 | 83 | ||
BC016494.1 | - | 6 - 7 | 967 | 4210859 | 83 | |
BC022151.1 | - | 6 - 7 | 960 | 5102845 | 83 | |
BC023894.1 | - | 6 - 7 | 974 | 5310164 | 83 | |
BC025822.1 | - | 6 - 7 | 984 | 5100453 | 83 | |
BC034831.1 | - | 6 - 7 | 966 | 5097081 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |