Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700007405.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007405.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007405.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007401.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007401.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007401.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007402.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007402.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007402.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007410.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007410.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007410.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007414.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007414.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007414.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007400.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007400.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007400.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007398.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007398.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007398.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007412.mm.1 |
TC0700000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700007412.mm.1 |
NM_025330.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007412.mm.1 |
NM_025330.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700007409.mm.1 |
TC0700000852.mm.1 |
MTA TC |