Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190795.1 | NP_001177724.1 | 9 - 10 | 1029 | 59 | |
NM_026409.4 | NP_080685.2 | 9 - 10 | 1029 | 59 | ||
GenBank mRNA | AK012782.1 | - | 9 - 10 | 1009 | 59 | |
AK017554.1 | - | 9 - 10 | 971 | 59 | ||
AK033741.1 | - | 9 - 10 | 1006 | 59 | ||
AK129402.1 | - | 9 - 10 | 1007 | 59 | ||
AK154422.1 | - | 9 - 10 | 1030 | 59 | ||
AK167149.1 | - | 8 - 9 | 3024 | 59 | ||
BC043052.1 | - | 9 - 10 | 979 | 59 | ||
BC117786.1 | - | 9 - 10 | 991 | 40092124 | 59 | |
BC117787.1 | - | 9 - 10 | 991 | 40092125 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |