Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011872.2 | NP_036002.1 | 3 - 4 | 295 | 78 | |
GenBank mRNA | AB008371.1 | - | 3 - 4 | 272 | 78 | |
AF124299.1 | - | 3 - 4 | 297 | 78 | ||
AK029477.1 | - | 3 - 4 | 295 | 78 | ||
AK077406.1 | - | 3 - 4 | 295 | 78 | ||
BC027823.1 | - | 3 - 4 | 456 | 5322691 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |