Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008861.3 | NP_032887.3 | 7 - 8 | 1884 | 112 | |
GenBank mRNA | AF014010.1 | - | 7 - 8 | 1787 | 112 | |
AK053502.1 | - | 7 - 8 | 1789 | 112 | ||
AK128961.1 | - | 8 - 9 | 1658 | 112 | ||
AK164673.1 | - | 7 - 8 | 1775 | 112 | ||
BC053058.1 | - | 7 - 8 | 1891 | 112 | ||
BC062969.1 | - | 7 - 8 | 1891 | 112 | ||
Y13278.1 | - | 7 - 8 | 1800 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |