Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198224.3 | NP_937867.2 | 1 - 2 | 174 | 102 | |
GenBank mRNA | AF544402.1 | - | 1 - 2 | 179 | 102 | |
BC089483.1 | - | 1 - 2 | 174 | 6772390 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |