Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016741.2 | NP_058021.1 | 12 - 13 | 1738 | 89 | |
GenBank mRNA | AK028191.1 | - | 12 - 13 | 1719 | 89 | |
AK033114.1 | - | 12 - 13 | 1663 | 89 | ||
AK154933.1 | - | 12 - 13 | 1730 | 89 | ||
AK158122.1 | - | 12 - 13 | 1663 | 89 | ||
AK169906.1 | - | 12 - 13 | 1714 | 89 | ||
AK212899.1 | - | 2 - 3 | 241 | 89 | ||
BC004656.1 | - | 12 - 13 | 1718 | 3498018 | 89 | |
U37799.1 | - | 12 - 13 | 1576 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |