Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178929.4 | NP_849260.2 | 1 - 2 | 105 | 75 | |
GenBank mRNA | BB618180.1 | - | 1 - 2 | 104 | 75 | |
BB871162.1 | - | 1 - 2 | 138 | 75 | ||
BC115642.1 | - | 1 - 2 | 96 | 40103018 | 75 | |
BC115643.1 | - | 1 - 2 | 93 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |