Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900005471.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005471.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005471.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005469.mm.1 |
NM_001164531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005469.mm.1 |
NM_001164531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005469.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005469.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005469.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005472.mm.1 |
NM_001164531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005472.mm.1 |
NM_001164531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005472.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005472.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005472.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005467.mm.1 |
NM_001164531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005467.mm.1 |
NM_001164531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005467.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005467.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005467.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005468.mm.1 |
NM_001164531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005468.mm.1 |
NM_001164531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005468.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005468.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005468.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005474.mm.1 |
NM_001164531.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005474.mm.1 |
NM_001164531.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005474.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005474.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005474.mm.1 |
TC1900000612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005470.mm.1 |
NM_177319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005470.mm.1 |
NM_177319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005470.mm.1 |
TC1900000612.mm.1 |
MTA TC |