Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008325.4 | NP_032351.2 | 7 - 8 | 1215 | 88 | |
NM_172997.1 | NP_766585.1 | 6 - 7 | 1074 | 88 | ||
GenBank mRNA | AK031414.1 | - | 6 - 7 | 869 | 88 | |
AK045330.1 | - | 5 - 6 | 702 | 88 | ||
AK155627.1 | - | 7 - 8 | 1216 | 88 | ||
AK169998.1 | - | 6 - 7 | 873 | 88 | ||
AK171124.1 | - | 2 - 3 | 226 | 88 | ||
BC057935.1 | - | 6 - 7 | 836 | 5327150 | 88 | |
L34111.1 | - | 6 - 7 | 789 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |