Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008325.4 | NP_032351.2 | 10 - 11 | 1824 | 105 | |
NM_172997.1 | NP_766585.1 | 9 - 10 | 1683 | 105 | ||
GenBank mRNA | AK031414.1 | - | 9 - 10 | 1478 | 105 | |
AK045330.1 | - | 8 - 9 | 1311 | 105 | ||
AK155627.1 | - | 10 - 11 | 1825 | 105 | ||
AK169998.1 | - | 9 - 10 | 1482 | 105 | ||
AK171124.1 | - | 5 - 6 | 835 | 105 | ||
BC057935.1 | - | 9 - 10 | 1445 | 5327150 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |