Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144846.5 | NP_659095.1 | 5 - 6 | 604 | 100 | |
GenBank mRNA | AK028062.1 | - | 5 - 6 | 605 | 100 | |
AK075613.1 | - | 6 - 7 | 549 | 100 | ||
AK205511.1 | - | 3 - 4 | 262 | 100 | ||
BC011343.1 | - | 5 - 6 | 422 | 4012217 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |