Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145156.1 | NP_660138.1 | 2 - 3 | 640 | 118 | |
GenBank mRNA | AF377994.1 | - | 2 - 3 | 458 | 118 | |
BC025908.1 | - | 2 - 3 | 640 | 5043992 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |